Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DFFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260683.1 | NP_001247612.1 | 205 | 61 | ||
XM_028844810.1 | XP_028700643.1 | 478 | 61 | |||
GenBank mRNA | JU333209.1 | - | 186 | 61 | ||
JU474525.1 | - | 205 | 61 | |||
JV644046.1 | - | 222 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |