Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DFFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260683.1 | NP_001247612.1 | 855 | 106 | ||
GenBank mRNA | JU333209.1 | - | 836 | 106 | ||
JU474525.1 | - | 855 | 106 | |||
JV644046.1 | - | 872 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |