Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028839122.1 | XP_028694955.1 | 4666 | 92 | ||
GenBank mRNA | JU334158.1 | - | 4148 | 92 | ||
JU475285.1 | - | 4155 | 92 | |||
JV046843.1 | - | 4153 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |