Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261149.1 | NP_001248078.1 | 1257 | 134 | ||
GenBank mRNA | JU477687.1 | - | 1257 | 134 | ||
JU477688.1 | - | 1245 | 134 | |||
JV048609.1 | - | 1239 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |