Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRMDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028826439.1 | XP_028682272.1 | 877 | 106 | ||
XM_028826442.1 | XP_028682275.1 | 1668 | 106 | |||
XM_028826443.1 | XP_028682276.1 | 721 | 106 | |||
GenBank mRNA | JV724786.1 | - | 397 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |