Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHCHD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266664.1 | NP_001253593.1 | 217 | 155 | ||
XM_015129926.2 | XP_014985412.2 | 317 | 155 | |||
XM_015129927.2 | XP_014985413.2 | 317 | 155 | |||
XM_015129928.2 | XP_014985414.2 | 523 | 155 | |||
XM_015129933.2 | XP_014985419.1 | 283 | 155 | |||
XM_028843420.1 | XP_028699253.1 | 416 | 155 | |||
XM_028843421.1 | XP_028699254.1 | 288 | 155 | |||
GenBank mRNA | JU336706.1 | - | 219 | 155 | ||
JV048439.1 | - | 217 | 155 | |||
JV637257.1 | - | 215 | 155 | |||
JV698622.1 | - | 227 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |