Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260651.1 | NP_001247580.1 | 129 | 58 | ||
GenBank mRNA | JV045378.1 | - | 129 | 58 | ||
JV636411.1 | - | 122 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |