Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP1LC3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261138.1 | NP_001248067.1 | 349 | 95 | ||
XM_028827301.1 | XP_028683134.1 | 978 | 95 | |||
XM_028827302.1 | XP_028683135.1 | 978 | 95 | |||
XM_028827303.1 | XP_028683136.1 | 957 | 95 | |||
GenBank mRNA | JU336751.1 | - | 345 | 95 | ||
JV045598.1 | - | 349 | 95 | |||
JV631144.1 | - | 352 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |