Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AOPEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015118085.2 | XP_014973571.2 | 2398 | 102 | ||
XM_015118087.2 | XP_014973573.2 | 2398 | 102 | |||
XM_015118088.2 | XP_014973574.2 | 2398 | 102 | |||
XM_015118090.2 | XP_014973576.2 | 2398 | 102 | |||
XM_015118091.2 | XP_014973577.2 | 2398 | 102 | |||
XM_015118094.2 | XP_014973580.2 | 2101 | 102 | |||
XM_028835409.1 | XP_028691242.1 | 2398 | 102 | |||
XM_028835410.1 | XP_028691243.1 | 2398 | 102 | |||
XM_028835411.1 | XP_028691244.1 | 2398 | 102 | |||
XM_028835412.1 | XP_028691245.1 | 2398 | 102 | |||
XM_028835413.1 | XP_028691246.1 | 2398 | 102 | |||
XM_028835414.1 | XP_028691247.1 | 2398 | 102 | |||
XM_028835416.1 | XP_028691249.1 | 2398 | 102 | |||
XM_028835417.1 | XP_028691250.1 | 2398 | 102 | |||
XM_028835418.1 | XP_028691251.1 | 2101 | 102 | |||
XM_028835420.1 | XP_028691253.1 | 2101 | 102 | |||
GenBank mRNA | JV073828.1 | - | 127 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |