Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RERG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261674.1 | NP_001248603.1 | 552 | 149 | ||
XM_015151102.2 | XP_015006588.1 | 859 | 149 | |||
GenBank mRNA | JU319806.1 | - | 552 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |