Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM241 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015121664.2 | XP_014977150.2 | 318 | 75 | ||
XM_015121665.2 | XP_014977151.2 | 318 | 75 | |||
XM_028837909.1 | XP_028693742.1 | 318 | 75 | |||
XM_028837911.1 | XP_028693744.1 | 318 | 75 | |||
XM_028837913.1 | XP_028693746.1 | 308 | 75 | |||
GenBank mRNA | JV663985.1 | - | 296 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |