Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXW11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261308.1 | NP_001248237.1 | 1331 | 100 | ||
XM_015141397.2 | XP_014996883.1 | 1495 | 100 | |||
XM_015141398.2 | XP_014996884.1 | 1645 | 100 | |||
XM_015141399.2 | XP_014996885.1 | 1655 | 100 | |||
XM_015141400.2 | XP_014996886.1 | 1552 | 100 | |||
XM_015141401.2 | XP_014996887.1 | 1543 | 100 | |||
GenBank mRNA | JU333627.1 | - | 1331 | 100 | ||
JU333628.1 | - | 1268 | 100 | |||
JU333629.1 | - | 1370 | 100 | |||
JU474886.1 | - | 1187 | 100 | |||
JV046558.1 | - | 1212 | 100 | |||
JV462163.1 | - | 1262 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |