Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS26B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257745.1 | NP_001244674.1 | 668 | 66 | ||
GenBank mRNA | JU320309.1 | - | 668 | 66 | ||
JU473342.1 | - | 531 | 66 | |||
JV044752.1 | - | 398 | 66 | |||
JV667667.1 | - | 681 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |