Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015127668.2 | XP_014983154.1 | 1045 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |