Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAP47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001087289.4 | XP_001087289.3 | 1192 | 159 | ||
XM_015114553.2 | XP_014970039.1 | 1205 | 159 | |||
XM_028826337.1 | XP_028682170.1 | 1285 | 159 | |||
XM_028826340.1 | XP_028682173.1 | 1102 | 159 | |||
XM_028826343.1 | XP_028682176.1 | 1199 | 159 | |||
GenBank mRNA | JU325109.1 | - | 1116 | 159 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |