Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001082781.4 | XP_001082781.2 | 925 | 96 | ||
XM_028846207.1 | XP_028702040.1 | 602 | 96 | |||
GenBank mRNA | JU329809.1 | - | 140 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |