Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257791.1 | NP_001244720.1 | 185 | 90 | ||
XM_015116590.2 | XP_014972076.1 | 314 | 90 | |||
GenBank mRNA | JU334049.1 | - | 185 | 90 | ||
JU475216.1 | - | 194 | 90 | |||
JV613509.1 | - | 195 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |