Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194184.2 | NP_001181113.1 | 304 | 107 | ||
XM_015116416.2 | XP_014971902.1 | 300 | 107 | |||
GenBank mRNA | JU332150.1 | - | 245 | 107 | ||
JV045421.1 | - | 269 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |