Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6PC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257733.1 | NP_001244662.1 | 509 | 107 | ||
GenBank mRNA | JU321767.1 | - | 509 | 107 | ||
JU473684.1 | - | 499 | 107 | |||
JV043843.1 | - | 510 | 107 | |||
JV637526.1 | - | 519 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |