Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REEP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194035.2 | NP_001180964.1 | 591 | 84 | ||
GenBank mRNA | JV048710.1 | - | 450 | 84 | ||
JV636458.1 | - | 428 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |