Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYND19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194731.2 | NP_001181660.1 | 770 | 60 | ||
GenBank mRNA | JU332828.1 | - | 579 | 60 | ||
JU474239.1 | - | 698 | 60 | |||
JV046132.1 | - | 570 | 60 | |||
JV601388.1 | - | 617 | 60 | |||
JV638766.1 | - | 733 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |