Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001104381.4 | XP_001104381.1 | 451 | 120 | ||
XM_015142784.2 | XP_014998270.1 | 432 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |