Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015131708.2 | XP_014987194.1 | 1285 | 123 | ||
XM_028843817.1 | XP_028699650.1 | 443 | 123 | |||
GenBank mRNA | JU333112.1 | - | 656 | 123 | ||
JU474444.1 | - | 675 | 123 | |||
JV572450.1 | - | 675 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |