Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPTC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194374.2 | NP_001181303.1 | 1127 | 83 | ||
GenBank mRNA | JU327311.1 | - | 759 | 83 | ||
JV643729.1 | - | 766 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |