Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015120293.2 | XP_014975779.1 | 2308 | 145 | ||
XM_015120294.2 | XP_014975780.1 | 1665 | 145 | |||
XM_015120298.2 | XP_014975784.1 | 1370 | 145 | |||
XM_015120299.2 | XP_014975785.1 | 1207 | 145 | |||
XM_028836778.1 | XP_028692611.1 | 1474 | 145 | |||
XM_028836779.1 | XP_028692612.1 | 1947 | 145 | |||
XM_028836780.1 | XP_028692613.1 | 1310 | 145 | |||
XM_028836781.1 | XP_028692614.1 | 1098 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |