Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194217.2 | NP_001181146.1 | 360 | 86 | ||
GenBank mRNA | JU320692.1 | - | 368 | 86 | ||
JU473285.1 | - | 585 | 86 | |||
JV048754.1 | - | 385 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |