Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC22B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261329.1 | NP_001248258.1 | 315 | 129 | ||
GenBank mRNA | JU337220.1 | - | 325 | 129 | ||
JU337221.1 | - | 325 | 129 | |||
JU477935.1 | - | 324 | 129 | |||
JV048776.1 | - | 315 | 129 | |||
JV666213.1 | - | 327 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |