Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VEGFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136099.1 | NP_001129571.1 | 818 | 75 | ||
GenBank mRNA | JV660540.1 | - | 681 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |