Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001105191.4 | XP_001105191.1 | 934 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |