Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDFI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257924.1 | NP_001244853.1 | 684 | 102 | ||
XM_015136323.2 | XP_014991809.1 | 598 | 102 | |||
XM_015136324.2 | XP_014991810.1 | 1556 | 102 | |||
XM_015136325.2 | XP_014991811.1 | 593 | 102 | |||
XM_015136326.2 | XP_014991812.1 | 530 | 102 | |||
GenBank mRNA | JU323225.1 | - | 684 | 102 | ||
JV602297.1 | - | 685 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |