Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGMAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257877.1 | NP_001244806.1 | 510 | 84 | ||
GenBank mRNA | JU320933.1 | - | 510 | 84 | ||
JU472376.1 | - | 450 | 84 | |||
JV044654.1 | - | 441 | 84 | |||
JV530655.1 | - | 515 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |