Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001094776.4 | XP_001094776.1 | 887 | 147 | ||
XM_015135494.2 | XP_014990980.1 | 871 | 147 | |||
XM_028847871.1 | XP_028703704.1 | 976 | 147 | |||
GenBank mRNA | JV527757.1 | - | 807 | 147 | ||
JV705242.1 | - | 737 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |