Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257480.1 | NP_001244409.1 | 340 | 147 | ||
XM_015132050.2 | XP_014987536.1 | 381 | 147 | |||
XM_015132051.2 | XP_014987537.1 | 376 | 147 | |||
XM_015132052.2 | XP_014987538.1 | 332 | 147 | |||
XM_028843422.1 | XP_028699255.1 | 460 | 147 | |||
XM_028843423.1 | XP_028699256.1 | 419 | 147 | |||
XM_028843424.1 | XP_028699257.1 | 261 | 147 | |||
GenBank mRNA | JU320326.1 | - | 340 | 147 | ||
JU475697.1 | - | 294 | 147 | |||
JV047145.1 | - | 240 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |