Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042638.2 | NP_001036103.2 | 418 | 112 | ||
XM_028832634.1 | XP_028688467.1 | 621 | 112 | |||
GenBank mRNA | CN647022.1 | - | 418 | 112 | ||
JU321360.1 | - | 518 | 112 | |||
JU473604.1 | - | 390 | 112 | |||
JV048086.1 | - | 406 | 112 | |||
JV556232.1 | - | 368 | 112 | |||
U93310.2 | - | 378 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |