Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032880.1 | NP_001028052.1 | 193 | 136 | ||
GenBank mRNA | AF449284.1 | - | 193 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |