Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P3H4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028836480.1 | XP_028692313.1 | 2113 | 103 | ||
XM_028836481.1 | XP_028692314.1 | 1533 | 103 | |||
GenBank mRNA | JU326395.1 | - | 840 | 103 | ||
JV045541.1 | - | 1068 | 103 | |||
JV454551.1 | - | 645 | 103 | |||
JV569158.1 | - | 677 | 103 | |||
JV705385.1 | - | 257 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |