Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGREF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015111945.2 | XP_014967431.2 | 381 | 60 | ||
XM_015111946.2 | XP_014967432.2 | 292 | 60 | |||
XM_015111947.2 | XP_014967433.2 | 381 | 60 | |||
GenBank mRNA | JU332573.1 | - | 135 | 60 | ||
JV569224.1 | - | 327 | 60 | |||
JV670900.1 | - | 310 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |