Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260866.1 | NP_001247795.1 | 309 | 153 | ||
GenBank mRNA | JU319794.1 | - | 378 | 153 | ||
JU473637.1 | - | 403 | 153 | |||
JV046000.1 | - | 309 | 153 | |||
JV529929.1 | - | 365 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |