Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266817.1 | NP_001253746.1 | 199 | 102 | ||
GenBank mRNA | JU323179.1 | - | 201 | 102 | ||
JU323242.1 | - | 183 | 102 | |||
JU476961.1 | - | 146 | 102 | |||
JV045068.1 | - | 199 | 102 | |||
JV703638.1 | - | 169 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |