Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265848.1 | NP_001252777.1 | 766 | 120 | ||
GenBank mRNA | JU326868.1 | - | 532 | 120 | ||
JU474115.1 | - | 799 | 120 | |||
JV046040.1 | - | 766 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |