Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST3GAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266341.1 | NP_001253270.1 | 1622 | 82 | ||
XM_028840536.1 | XP_028696369.1 | 1723 | 82 | |||
GenBank mRNA | JU332128.1 | - | 1492 | 82 | ||
JU470852.1 | - | 1631 | 82 | |||
JV044250.1 | - | 1622 | 82 | |||
JV567663.1 | - | 1679 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |