Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028841165.1 | XP_028696998.1 | 700 | 143 | ||
XM_028841166.1 | XP_028696999.1 | 470 | 143 | |||
XM_028841167.1 | XP_028697000.1 | 326 | 143 | |||
XM_028841168.1 | XP_028697001.1 | 795 | 143 | |||
XM_028841169.1 | XP_028697002.1 | 331 | 143 | |||
GenBank mRNA | JU323934.1 | - | 425 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |