Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001095296.4 | XP_001095296.2 | 1285 | 85 | ||
GenBank mRNA | JV044396.1 | - | 795 | 85 | ||
JV663997.1 | - | 740 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |