Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR1H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194697.1 | NP_001181626.1 | 739 | 74 | ||
XM_015124413.2 | XP_014979899.1 | 505 | 74 | |||
XM_015124414.2 | XP_014979900.1 | 685 | 74 | |||
GenBank mRNA | JU336224.1 | - | 676 | 74 | ||
JU472352.1 | - | 597 | 74 | |||
JV045753.1 | - | 581 | 74 | |||
JV556720.1 | - | 687 | 74 | |||
JV627735.1 | - | 708 | 74 |