Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001095865.4 | XP_001095865.1 | 642 | 149 | ||
XM_002800178.3 | XP_002800224.1 | 749 | 149 | |||
XM_015116797.2 | XP_014972283.1 | 635 | 149 | |||
XM_028834805.1 | XP_028690638.1 | 589 | 149 | |||
GenBank mRNA | JU336290.1 | - | 250 | 149 | ||
JU493006.1 | - | 653 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |