Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSIG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001099264.2 | XP_001099264.2 | 535 | 97 | ||
XM_015127593.2 | XP_014983079.1 | 535 | 97 | |||
XM_015127594.1 | XP_014983080.1 | 535 | 97 | |||
XM_015127595.2 | XP_014983081.1 | 535 | 97 | |||
XM_028841902.1 | XP_028697735.1 | 535 | 97 | |||
XM_028841903.1 | XP_028697736.1 | 535 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |