Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015133750.2 | XP_014989236.1 | 1199 | 135 | ||
XM_028845883.1 | XP_028701716.1 | 835 | 135 | |||
GenBank mRNA | JU320866.1 | - | 1144 | 135 | ||
JV044565.1 | - | 1216 | 135 | |||
JV044566.1 | - | 1216 | 135 | |||
JV044567.1 | - | 1216 | 135 | |||
JV044568.1 | - | 1216 | 135 | |||
JV493533.1 | - | 1027 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |