Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265874.1 | NP_001252803.1 | 667 | 106 | ||
XM_015139520.2 | XP_014995006.1 | 710 | 106 | |||
GenBank mRNA | JU333948.1 | - | 586 | 106 | ||
JU475132.1 | - | 465 | 106 | |||
JV046732.1 | - | 667 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |