Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261196.1 | NP_001248125.1 | 118 | 85 | ||
XM_028849099.1 | XP_028704932.1 | 148 | 85 | |||
GenBank mRNA | JU333728.1 | - | 112 | 85 | ||
JU474973.1 | - | 118 | 85 | |||
JV710937.1 | - | 148 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |