Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261097.1 | NP_001248026.1 | 159 | 92 | ||
XM_015124420.2 | XP_014979906.2 | 352 | 92 | |||
GenBank mRNA | JU333851.1 | - | 159 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |